Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010593.2 | NP_034723.1 | 5 - 6 | 1069 | 98 | |
GenBank mRNA | AK145895.1 | - | 5 - 6 | 1054 | 98 | |
AK170934.1 | - | 5 - 6 | 1042 | 98 | ||
AK171501.1 | - | 1193 | 98 | |||
BC040757.1 | - | 5 - 6 | 1018 | 4216772 | 98 | |
BC094461.1 | - | 5 - 6 | 1022 | 4459882 | 98 | |
M90365.1 | - | 3 - 4 | 534 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |