Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013495.2 | NP_038523.2 | 5 - 6 | 638 | 60 | |
GenBank mRNA | AF017175.1 | - | 526 | 60 | ||
AK050213.1 | - | 5 - 6 | 931 | 60 | ||
AK077454.1 | - | 5 - 6 | 592 | 60 | ||
AK128925.1 | - | 5 - 6 | 592 | 60 | ||
AK133472.1 | - | 5 - 6 | 593 | 60 | ||
AK136487.1 | - | 5 - 6 | 592 | 60 | ||
AK147770.1 | - | 5 - 6 | 593 | 60 | ||
AK149688.1 | - | 5 - 6 | 589 | 60 | ||
BC038395.1 | - | 4 - 5 | 573 | 2649352 | 60 | |
BC046383.1 | - | 4 - 5 | 573 | 4160676 | 60 | |
BC054791.1 | - | 5 - 6 | 619 | 6832731 | 60 | |
D49472.1 | - | 4 - 5 | 565 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |