Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037877.3 | NP_001032966.1 | 11 - 12 | 1379 | 72 | |
NM_001037878.3 | NP_001032967.1 | 11 - 12 | 1379 | 72 | ||
NM_001286362.1 | NP_001273291.1 | 10 - 11 | 1304 | 72 | ||
NM_001286363.1 | NP_001273292.1 | 10 - 11 | 1304 | 72 | ||
NM_025804.3 | NP_080080.2 | 11 - 12 | 1379 | 72 | ||
GenBank mRNA | AK003180.1 | - | 11 - 12 | 1267 | 72 | |
AK013754.1 | - | 11 - 12 | 1068 | 72 | ||
AK045397.1 | - | 10 - 11 | 1232 | 72 | ||
AK050274.1 | - | 11 - 12 | 1307 | 72 | ||
AK076151.1 | - | 5 - 6 | 466 | 72 | ||
AK088360.1 | - | 11 - 12 | 1267 | 72 | ||
AK089964.1 | - | 11 - 12 | 1268 | 72 | ||
AK122430.1 | - | 11 - 12 | 1234 | 72 | ||
AK148082.1 | - | 11 - 12 | 1241 | 72 | ||
AK150232.1 | - | 11 - 12 | 1266 | 72 | ||
AK150918.1 | - | 11 - 12 | 72 | |||
AK150942.1 | - | 11 - 12 | 72 | |||
AK151143.1 | - | 11 - 12 | 72 | |||
AK151558.1 | - | 11 - 12 | 1260 | 72 | ||
AK152178.1 | - | 11 - 12 | 72 | |||
AK152494.1 | - | 11 - 12 | 72 | |||
AK161627.1 | - | 11 - 12 | 1255 | 72 | ||
AK167837.1 | - | 11 - 12 | 1253 | 72 | ||
AK168167.1 | - | 11 - 12 | 1253 | 72 | ||
BC016130.1 | - | 11 - 12 | 1241 | 72 | ||
BC025071.1 | - | 11 - 12 | 1240 | 3498003 | 72 | |
BC046768.1 | - | 11 - 12 | 1337 | 5707312 | 72 | |
EU624323.1 | - | 10 - 11 | 1142 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |