Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190453.1 | NP_001177382.1 | 5 - 6 | 509 | 76 | |
NM_001190454.1 | NP_001177383.1 | 4 - 5 | 518 | 76 | ||
NM_027151.2 | NP_081427.1 | 4 - 5 | 503 | 76 | ||
GenBank mRNA | AK144464.1 | - | 4 - 5 | 379 | 76 | |
AK144505.1 | - | 4 - 5 | 379 | 76 | ||
AK164021.1 | - | 4 - 5 | 380 | 76 | ||
BC004613.1 | - | 4 - 5 | 366 | 3585310 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |