Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028665.4 | NP_082941.2 | 2 - 3 | 1173 | 69 | |
GenBank mRNA | AK029511.1 | - | 2 - 3 | 1175 | 69 | |
AK029888.1 | - | 2 - 3 | 1134 | 69 | ||
AK033599.1 | - | 2 - 3 | 1162 | 69 | ||
AK048925.1 | - | 2 - 3 | 1067 | 69 | ||
AK133339.1 | - | 1134 | 69 | |||
BC030885.1 | - | 2 - 3 | 802 | 69 | ||
BC100433.1 | - | 2 - 3 | 1167 | 69 | ||
BC145456.1 | - | 2 - 3 | 291 | 9054017 | 69 | |
BC145457.1 | - | 2 - 3 | 291 | 69 | ||
BC145458.1 | - | 2 - 3 | 291 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |