Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028799.2 | NP_083075.1 | 9 - 10 | 1442 | 133 | |
GenBank mRNA | AK009196.2 | - | 9 - 10 | 1442 | 133 | |
AK037074.1 | - | 9 - 10 | 1503 | 133 | ||
AK132482.1 | - | 9 - 10 | 1525 | 133 | ||
BC116266.1 | - | 9 - 10 | 1425 | 40094000 | 133 | |
BC116267.1 | - | 9 - 10 | 1425 | 40094002 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |