Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172282.2 | NP_758486.1 | 8 - 9 | 1793 | 85 | |
GenBank mRNA | AK033499.1 | - | 8 - 9 | 1554 | 85 | |
AK046065.1 | - | 8 - 9 | 1580 | 85 | ||
AK049974.1 | - | 8 - 9 | 1549 | 85 | ||
AK088866.1 | - | 8 - 9 | 1577 | 85 | ||
AK148252.1 | - | 8 - 9 | 1596 | 85 | ||
BC026890.1 | - | 1 - 2 | 66 | 85 | ||
BC039955.1 | - | 8 - 9 | 1539 | 4236297 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |