Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spire2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172287.2 | NP_758491.1 | 1 - 2 | 306 | 96 | |
GenBank mRNA | BC049152.1 | - | 1 - 2 | 306 | 6311485 | 96 |
BC054072.1 | - | 1 - 2 | 306 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |