Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136471.1 | NP_001129943.1 | 5 - 6 | 597 | 97 | |
NM_172297.1 | NP_758501.2 | 4 - 5 | 614 | 97 | ||
GenBank mRNA | AK134892.1 | - | 5 - 6 | 583 | 97 | |
BC021915.1 | - | 4 - 5 | 614 | 4527178 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |