Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163755.1 | NP_001157227.1 | 3 - 4 | 2420 | 70 | |
NM_001163757.1 | NP_001157229.1 | 3 - 4 | 2420 | 70 | ||
NM_001163758.1 | NP_001157230.1 | 3 - 4 | 2401 | 70 | ||
NM_172446.3 | NP_766034.2 | 3 - 4 | 2473 | 70 | ||
GenBank mRNA | AB185113.1 | - | 3 - 4 | 2423 | 70 | |
AK029916.1 | - | 3 - 4 | 2404 | 70 | ||
AK049035.1 | - | 3 - 4 | 2474 | 70 | ||
AK134840.1 | - | 3 - 4 | 2412 | 70 | ||
BC138063.1 | - | 3 - 4 | 2377 | 8861082 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |