Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001295.2 | NP_001001295.2 | 8 - 9 | 1292 | 55 | |
NM_001177784.1 | NP_001171255.1 | 8 - 9 | 1253 | 55 | ||
NM_172519.3 | NP_766107.1 | 8 - 9 | 1196 | 55 | ||
GenBank mRNA | AK029974.1 | - | 8 - 9 | 1197 | 55 | |
AK032451.1 | - | 8 - 9 | 1201 | 55 | ||
AK077031.1 | - | 8 - 9 | 1256 | 55 | ||
AK150851.1 | - | 8 - 9 | 1242 | 55 | ||
AK151780.1 | - | 8 - 9 | 1242 | 55 | ||
AK153433.1 | - | 8 - 9 | 1242 | 55 | ||
AK173310.1 | - | 8 - 9 | 1226 | 55 | ||
BC056939.1 | - | 8 - 9 | 1225 | 5708685 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |