Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172526.3 | NP_766114.1 | 7 - 8 | 1367 | 64 | |
GenBank mRNA | AK039884.1 | - | 7 - 8 | 1368 | 64 | |
AK076743.1 | - | 2 - 3 | 183 | 64 | ||
BC055114.1 | - | 2 - 3 | 182 | 64 | ||
BC147554.1 | - | 6 - 7 | 1071 | 9056545 | 64 | |
BC147555.1 | - | 6 - 7 | 1071 | 9056546 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |