Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146048.1 | NP_001139520.1 | 9 - 10 | 1134 | 128 | |
NM_172528.3 | NP_766116.3 | 9 - 10 | 782 | 128 | ||
GenBank mRNA | AK051043.1 | - | 9 - 10 | 1133 | 128 | |
AK082721.1 | - | 9 - 10 | 783 | 128 | ||
AK147075.1 | - | 9 - 10 | 1134 | 128 | ||
AK220166.1 | - | 9 - 10 | 1152 | 128 | ||
BC022613.1 | - | 1 - 2 | 76 | 128 | ||
BC046591.1 | - | 9 - 10 | 1121 | 128 | ||
BC087542.1 | - | 9 - 10 | 981 | 4934980 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |