Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Engase | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172573.2 | NP_766161.1 | 6 - 7 | 874 | 60 | |
GenBank mRNA | AK048667.1 | - | 6 - 7 | 876 | 60 | |
AK084254.1 | - | 6 - 7 | 1007 | 60 | ||
AK167142.1 | - | 6 - 7 | 846 | 60 | ||
AK171813.1 | - | 1 - 2 | 198 | 60 | ||
BC060640.1 | - | 5 - 6 | 698 | 60 | ||
BC120923.1 | - | 6 - 7 | 872 | 60 | ||
BC120924.1 | - | 6 - 7 | 876 | 40097903 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |