Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172582.3 | NP_766170.2 | 10 - 11 | 1315 | 73 | |
GenBank mRNA | AK077863.1 | - | 10 - 11 | 1239 | 73 | |
AK131889.1 | - | 10 - 11 | 1277 | 73 | ||
AK167380.1 | - | 10 - 11 | 1243 | 73 | ||
AK170311.1 | - | 9 - 10 | 1780 | 73 | ||
AK191776.1 | - | 1 - 2 | 85 | 73 | ||
BC024135.1 | - | 10 - 11 | 1216 | 73 | ||
BC051055.1 | - | 10 - 11 | 1231 | 73 | ||
BC057914.1 | - | 7 - 8 | 979 | 73 | ||
BC088736.1 | - | 6 - 7 | 2564 | 73 | ||
BC106183.1 | - | 10 - 11 | 1299 | 73 | ||
BC132168.1 | - | 10 - 11 | 1315 | 40130445 | 73 | |
BC132170.1 | - | 10 - 11 | 1315 | 40130447 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |