Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205353.1 | NP_001192282.1 | 12 - 13 | 1277 | 59 | |
NM_172611.4 | NP_766199.1 | 12 - 13 | 1179 | 59 | ||
GenBank mRNA | AK036832.1 | - | 12 - 13 | 1168 | 59 | |
AK142266.1 | - | 12 - 13 | 1162 | 59 | ||
BC021523.1 | - | 8 - 9 | 553 | 59 | ||
BC085148.1 | - | 12 - 13 | 1235 | 59 | ||
BC139228.1 | - | 12 - 13 | 1168 | 8862250 | 59 | |
BC139229.1 | - | 12 - 13 | 1168 | 8862251 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |