Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Treml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033922.2 | NP_001029094.1 | 3 - 4 | 545 | 66 | |
NM_172623.2 | NP_766211.2 | 3 - 4 | 557 | 66 | ||
GenBank mRNA | AK077228.1 | - | 3 - 4 | 557 | 66 | |
AK156085.1 | - | 3 - 4 | 560 | 66 | ||
AY557629.1 | - | 3 - 4 | 493 | 66 | ||
BC117091.1 | - | 3 - 4 | 532 | 40126410 | 66 | |
BC137666.1 | - | 3 - 4 | 532 | 8860685 | 66 | |
DQ087185.1 | - | 3 - 4 | 479 | 66 | ||
DQ087186.1 | - | 3 - 4 | 479 | 66 | ||
DQ186654.1 | - | 3 - 4 | 499 | 66 | ||
DQ186655.1 | - | 3 - 4 | 511 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |