Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038619.1 | NP_001033708.1 | 10 - 11 | 1541 | 88 | |
NM_172646.2 | NP_766234.1 | 10 - 11 | 1545 | 88 | ||
GenBank mRNA | AK036199.1 | - | 10 - 11 | 1506 | 88 | |
AK049724.1 | - | 10 - 11 | 1546 | 88 | ||
AK082698.1 | - | 10 - 11 | 1485 | 88 | ||
AK122379.1 | - | 1 - 2 | 155 | 88 | ||
AK147678.1 | - | 10 - 11 | 1485 | 88 | ||
BC141143.1 | - | 10 - 11 | 1389 | 9055755 | 88 | |
BC141144.1 | - | 10 - 11 | 1389 | 9055756 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |