Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172709.3 | NP_766297.2 | 1 - 2 | 425 | 80 | |
GenBank mRNA | AF548337.1 | - | 1 - 2 | 425 | 80 | |
AK040792.1 | - | 1 - 2 | 412 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |