Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172735.2 | NP_766323.1 | 7 - 8 | 1018 | 68 | |
GenBank mRNA | AJ537495.1 | - | 7 - 8 | 1010 | 68 | |
AK088527.1 | - | 7 - 8 | 1019 | 68 | ||
AK146228.1 | - | 7 - 8 | 1033 | 68 | ||
AK151694.1 | - | 7 - 8 | 1014 | 68 | ||
AK167992.1 | - | 7 - 8 | 1033 | 68 | ||
AK168071.1 | - | 7 - 8 | 1028 | 68 | ||
BC024560.1 | - | 1 - 2 | 34 | 68 | ||
BC037445.1 | - | 7 - 8 | 972 | 68 | ||
BC050141.1 | - | 7 - 8 | 1011 | 30011212 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |