Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172757.3 | NP_766345.3 | 1 - 2 | 273 | 93 | |
GenBank mRNA | AK047782.1 | - | 1 - 2 | 304 | 93 | |
AK052635.1 | - | 1 - 2 | 300 | 93 | ||
AK134510.1 | - | 1 - 2 | 93 | |||
AK136424.1 | - | 1 - 2 | 282 | 93 | ||
AK140522.1 | - | 1 - 2 | 291 | 93 | ||
AK163715.1 | - | 1 - 2 | 260 | 93 | ||
BC065060.1 | - | 1 - 2 | 273 | 6402217 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |