Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172758.4 | NP_766346.1 | 9 - 10 | 1281 | 57 | |
GenBank mRNA | AK052548.1 | - | 9 - 10 | 1282 | 57 | |
AK171137.1 | - | 9 - 10 | 1289 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Mus musculus Probe spans exons |