Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400003262.mm.1 |
NM_172808.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003262.mm.1 |
NM_172808.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003262.mm.1 |
TC1400000367.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003263.mm.1 |
NM_172808.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003263.mm.1 |
NM_172808.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003263.mm.1 |
TC1400000367.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003265.mm.1 |
NM_172808.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003265.mm.1 |
NM_172808.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003265.mm.1 |
TC1400000367.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003261.mm.1 |
NM_172808.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003261.mm.1 |
NM_172808.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003261.mm.1 |
TC1400000367.mm.1 |
MTA TC |