Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172815.3 | NP_766403.1 | 1 - 2 | 86 | ||
GenBank mRNA | AK087485.1 | - | 1 - 2 | 86 | ||
BC052844.1 | - | 1 - 2 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |