Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172845.2 | NP_766433.1 | 5 - 6 | 75 | ||
GenBank mRNA | AK028364.1 | - | 75 | |||
AK076295.1 | - | 4 - 5 | 75 | |||
AK083534.1 | - | 5 - 6 | 75 | |||
AK156429.1 | - | 3 - 4 | 75 | |||
AK157287.1 | - | 75 | ||||
AK164900.1 | - | 5 - 6 | 75 | |||
AK173001.1 | - | 1847 | 75 | |||
BC027773.1 | - | 5 - 6 | 1603 | 5345809 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |