Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793481 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793476 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793480 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793477 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793479 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793478 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793482 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P618132 |
BC067076.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P618132 |
AK077822.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P618132 |
NM_172865.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P618132 |
BC067076.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P618132 |
AK077822.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P618132 |
NM_172865.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
278532 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
573848 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455573 |
AK082041.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455573 |
BC066105.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455573 |
NM_001110276.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729162 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
559678 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616228 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
827614 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
286481 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
374087 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
908478 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
174122 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
946370 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364955 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
306812 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
441338 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
26623 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
162561 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135622 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P137625 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P618132 |
BC067076.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P618132 |
AK077822.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P618132 |
NM_172865.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P618132 |
BC067076.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P618132 |
AK077822.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P618132 |
NM_172865.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793501 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793477 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793513 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793495 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793497 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793491 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793506 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793482 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793503 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793504 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793470 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793507 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793488 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793499 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793462 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793489 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793472 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793478 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793461 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793509 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793474 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793463 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793459 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793480 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793467 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793493 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793469 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793511 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793465 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793514 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793487 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22793486 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05183147 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05183171 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135622 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P137625 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P239884 |
BC067076.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P239884 |
AK030141.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P239884 |
NM_172865.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P234214 |
AK083774.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P618132 |
BC067076.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P618132 |
AK077822.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P618132 |
NM_172865.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P618132 |
BC067076.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P618132 |
AK077822.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P618132 |
NM_172865.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121687 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135622 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P137625 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03720239 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03778069 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03288278 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03720240 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793482 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793476 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793478 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793477 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793480 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793479 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22793481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793400 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793319 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793448 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793329 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793325 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793405 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793422 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793362 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793403 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793322 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793530 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793471 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793385 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793331 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793343 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793327 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793517 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793317 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793477 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793366 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793392 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793381 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793341 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793469 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793458 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793506 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793426 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793507 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793503 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793465 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793491 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793364 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793474 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793526 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22793486 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793465 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793458 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793451 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793487 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03754934 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03426971 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793524 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793363 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793514 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03857152 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793350 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03212046 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03269751 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793470 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793431 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793496 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793486 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03754912 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793495 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793442 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793509 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793504 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793335 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793366 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793474 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03605371 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793515 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793355 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793354 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793469 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793448 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793480 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793362 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793325 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03364727 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793517 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793497 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03364729 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793321 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793507 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793491 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793483 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793439 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793503 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03426964 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03754916 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793501 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793358 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793521 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793436 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03754920 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793323 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03501308 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793329 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793361 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793472 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793322 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793513 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793493 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793523 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03475510 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793477 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04076850 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793489 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793434 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793471 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03501305 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793364 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793351 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793454 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793327 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03754924 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793446 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793478 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793499 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03801151 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793462 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793506 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793331 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793425 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793526 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793456 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793441 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793516 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793450 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793320 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793511 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22793359 |
- |
- |