Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172866.3 | NP_766454.1 | 2 - 3 | 408 | 70 | |
GenBank mRNA | AK089288.1 | - | 2 - 3 | 191 | 70 | |
AK129103.1 | - | 2 - 3 | 194 | 70 | ||
AK170117.1 | - | 2 - 3 | 205 | 70 | ||
BC086614.1 | - | 2 - 3 | 214 | 30533256 | 70 | |
CJ065220.1 | - | 2 - 3 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |