Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172866.3 | NP_766454.1 | 7 - 8 | 1051 | 79 | |
GenBank mRNA | AK089288.1 | - | 7 - 8 | 834 | 79 | |
AK129103.1 | - | 7 - 8 | 837 | 79 | ||
AK170117.1 | - | 7 - 8 | 848 | 79 | ||
BC086614.1 | - | 7 - 8 | 857 | 30533256 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |