Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Podn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285956.1 | NP_001272885.1 | 8 - 9 | 1216 | 87 | |
NM_001285958.1 | NP_001272887.1 | 8 - 9 | 1112 | 87 | ||
NM_172874.3 | NP_766462.2 | 8 - 9 | 1138 | 87 | ||
GenBank mRNA | AK034987.1 | - | 7 - 8 | 1669 | 87 | |
AK161018.1 | - | 8 - 9 | 1002 | 87 | ||
AY313606.1 | - | 8 - 9 | 1138 | 87 | ||
BC052487.1 | - | 8 - 9 | 963 | 87 | ||
BC064034.1 | - | 3 - 4 | 300 | 87 | ||
BC094340.1 | - | 8 - 9 | 1216 | 30609151 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |