Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172883.2 | NP_766471.1 | 5 - 6 | 763 | 81 | |
GenBank mRNA | AK029021.1 | - | 5 - 6 | 763 | 81 | |
AK087226.1 | - | 5 - 6 | 973 | 81 | ||
AK171069.1 | - | 4 - 5 | 3395 | 81 | ||
BC094512.1 | - | 3 - 4 | 375 | 2616131 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |