Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173180.3 | NP_775272.1 | 7 - 8 | 906 | 71 | |
GenBank mRNA | AK004549.1 | - | 7 - 8 | 906 | 71 | |
AK032081.1 | - | 7 - 8 | 905 | 71 | ||
AK075965.1 | - | 7 - 8 | 71 | |||
AK077218.1 | - | 7 - 8 | 907 | 71 | ||
AK147110.1 | - | 7 - 8 | 909 | 71 | ||
AK150118.1 | - | 7 - 8 | 907 | 71 | ||
AK158990.1 | - | 7 - 8 | 874 | 71 | ||
AK161291.1 | - | 7 - 8 | 907 | 71 | ||
AK167975.1 | - | 7 - 8 | 905 | 71 | ||
AK169325.1 | - | 7 - 8 | 901 | 71 | ||
AK169342.1 | - | 7 - 8 | 901 | 71 | ||
AK196483.1 | - | 1 - 2 | 38 | 71 | ||
BC010810.1 | - | 7 - 8 | 891 | 4241697 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |