Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173400.2 | NP_775576.2 | 1 - 2 | 337 | 69 | |
GenBank mRNA | AK040101.1 | - | 1 - 2 | 340 | 69 | |
AK051016.1 | - | 1 - 2 | 357 | 69 | ||
AK077681.1 | - | 1 - 2 | 357 | 69 | ||
AK169222.1 | - | 1 - 2 | 341 | 69 | ||
AK173206.1 | - | 1 - 2 | 311 | 69 | ||
BC068237.1 | - | 1 - 2 | 337 | 30550558 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |