Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175543.3 | NP_780752.1 | 12 - 13 | 1553 | 74 | |
GenBank mRNA | AK046734.1 | - | 12 - 13 | 1555 | 74 | |
AK122559.1 | - | 4 - 5 | 389 | 74 | ||
AK147219.1 | - | 12 - 13 | 1624 | 74 | ||
AK147344.1 | - | 12 - 13 | 1749 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |