Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007586.1 | NP_031612.1 | 4 - 5 | 417 | 60 | |
GenBank mRNA | AK032171.1 | - | 423 | 60 | ||
BC017646.1 | - | 4 - 5 | 417 | 4527074 | 60 | |
X73985.1 | - | 2 - 3 | 106 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |