Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008323.1 | NP_032349.1 | 10 - 11 | 965 | 91 | |
GenBank mRNA | AJ748655.1 | - | 10 - 11 | 917 | 91 | |
AK003109.1 | - | 1 - 2 | 53 | 91 | ||
AK156309.1 | - | 10 - 11 | 963 | 91 | ||
AK166925.1 | - | 9 - 10 | 922 | 91 | ||
AK167131.1 | - | 7 - 8 | 1683 | 91 | ||
AK168526.1 | - | 10 - 11 | 963 | 91 | ||
BC085179.1 | - | 10 - 11 | 938 | 30699000 | 91 | |
BC119170.1 | - | 10 - 11 | 948 | 8733919 | 91 | |
BC119172.1 | - | 10 - 11 | 948 | 8733921 | 91 | |
U68564.1 | - | 10 - 11 | 965 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |