Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600014085.mm.1 |
NM_001159394.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014085.mm.1 |
NM_001159394.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014085.mm.1 |
NM_030612.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014085.mm.1 |
NM_030612.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014085.mm.1 |
TC1600001742.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014089.mm.1 |
NM_001159395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014089.mm.1 |
NM_001159395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014089.mm.1 |
NM_030612.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014089.mm.1 |
NM_030612.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014089.mm.1 |
TC1600001742.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600014090.mm.1 |
NM_001159395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014090.mm.1 |
NM_001159395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014090.mm.1 |
NM_030612.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014090.mm.1 |
NM_030612.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600014090.mm.1 |
TC1600001742.mm.1 |
MTA TC |