Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172157.1 | NP_001165628.1 | 12 - 13 | 1518 | 63 | |
NM_153530.2 | NP_705758.1 | 11 - 12 | 1476 | 63 | ||
GenBank mRNA | AK029652.1 | - | 12 - 13 | 1532 | 63 | |
AK031180.1 | - | 12 - 13 | 1501 | 63 | ||
BC036177.1 | - | 11 - 12 | 1437 | 5004341 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |