Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sqrdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162503.1 | NP_001155975.1 | 7 - 8 | 1150 | 133 | |
NM_021507.5 | NP_067482.4 | 7 - 8 | 1469 | 133 | ||
NR_027888.1 | 7 - 8 | 1105 | 133 | |||
GenBank mRNA | AF174535.1 | - | 7 - 8 | 1469 | 133 | |
AK008290.1 | - | 7 - 8 | 1116 | 133 | ||
AK075598.1 | - | 7 - 8 | 1151 | 133 | ||
AK149948.1 | - | 7 - 8 | 1121 | 133 | ||
BC011153.1 | - | 6 - 7 | 1088 | 4196280 | 133 | |
BC028247.1 | - | 7 - 8 | 1137 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |