Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175114.3 | NP_780323.2 | 4 - 5 | 625 | 68 | |
GenBank mRNA | AK029123.1 | - | 5 - 6 | 1023 | 68 | |
AK041436.1 | - | 5 - 6 | 993 | 68 | ||
AK122426.1 | - | 5 - 6 | 715 | 68 | ||
BC046285.1 | - | 4 - 5 | 615 | 68 | ||
BC058971.1 | - | 4 - 5 | 567 | 6832507 | 68 | |
BC116262.1 | - | 4 - 5 | 592 | 68 | ||
BC116263.1 | - | 4 - 5 | 592 | 68 | ||
BC129961.1 | - | 4 - 5 | 592 | 68 | ||
BC129962.1 | - | 4 - 5 | 592 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |