Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291002.1 | NP_001277931.1 | 3 - 4 | 377 | 85 | |
NM_175154.3 | NP_780363.1 | 3 - 4 | 483 | 85 | ||
GenBank mRNA | AK053002.1 | - | 3 - 4 | 333 | 85 | |
AK082521.1 | - | 3 - 4 | 374 | 85 | ||
AK152851.1 | - | 1 - 2 | 67 | 85 | ||
AK154762.1 | - | 3 - 4 | 332 | 85 | ||
AK171939.1 | - | 3 - 4 | 360 | 85 | ||
BC079843.1 | - | 3 - 4 | 347 | 5690056 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |