Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293784.1 | NP_001280713.1 | 8 - 9 | 1041 | 79 | |
NM_175188.4 | NP_780397.2 | 7 - 8 | 988 | 79 | ||
GenBank mRNA | AK036489.1 | - | 3 - 4 | 552 | 79 | |
AK086052.1 | - | 8 - 9 | 1000 | 79 | ||
AK152592.1 | - | 3 - 4 | 553 | 79 | ||
BC066008.1 | - | 7 - 8 | 988 | 5703753 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |