Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291069.1 | NP_001277998.1 | 4 - 5 | 1023 | 70 | |
NM_175193.6 | NP_780402.1 | 4 - 5 | 1023 | 70 | ||
GenBank mRNA | AK033730.1 | - | 4 - 5 | 1010 | 70 | |
AK048434.1 | - | 5 - 6 | 934 | 70 | ||
AK078928.1 | - | 4 - 5 | 1023 | 70 | ||
AK149286.1 | - | 4 - 5 | 983 | 70 | ||
BC086642.1 | - | 4 - 5 | 855 | 6836803 | 70 | |
BC089354.1 | - | 4 - 5 | 905 | 6837211 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |