Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252494.1 | NP_001239423.1 | 7 - 8 | 829 | 104 | |
NM_001252496.1 | NP_001239425.1 | 7 - 8 | 829 | 104 | ||
NM_001252497.1 | NP_001239426.1 | 2 - 3 | 1087 | 104 | ||
NM_001252498.1 | NP_001239427.1 | 2 - 3 | 1087 | 104 | ||
NM_175258.4 | NP_780467.2 | 7 - 8 | 829 | 104 | ||
GenBank mRNA | AK045540.1 | - | 7 - 8 | 771 | 104 | |
AK049228.1 | - | 7 - 8 | 785 | 104 | ||
AK049669.1 | - | 2 - 3 | 1088 | 104 | ||
AK164343.1 | - | 2 - 3 | 1060 | 104 | ||
AK167055.1 | - | 7 - 8 | 830 | 104 | ||
BC141182.1 | - | 7 - 8 | 728 | 9055794 | 104 | |
BC145503.1 | - | 7 - 8 | 728 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |