Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2151091 |
BC037059.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2151091 |
BC075672.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2151091 |
BC096033.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2151091 |
NM_175306.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2151091 |
AK029356.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
626306 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455408 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
211052 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
641931 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
345126 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428782 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
884499 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
181910 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
240482 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
404324 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798852 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497084 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
170311 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491438 |
AK003656.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491438 |
AK030589.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491438 |
AK049110.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491438 |
AK143891.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491438 |
AK154968.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491438 |
AK173121.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491438 |
AY052398.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491438 |
BC023397.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
491438 |
NM_029098.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
451860 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
806534 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
617251 |
AK017263.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
924931 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760638 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
745774 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
471857 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882532 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
712190 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330814 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
271464 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
47063 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
174692 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
518113 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402412 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
326411 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
462968 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
508433 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
544802 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
660480 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
348553 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
340512 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
736309 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385253 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
828194 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
117869 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
96285 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
121791 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
15712 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
63632 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
163778 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
158120 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2151091 |
BC037059.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2151091 |
BC075672.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2151091 |
BC096033.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2151091 |
NM_175306.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2151091 |
AK029356.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P242526 |
BC075672.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P242526 |
BC096033.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P242526 |
NM_175306.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P242526 |
AK049209.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P242526 |
AK220496.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323221 |
BC037059.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323221 |
BC075672.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323221 |
BC096033.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323221 |
NM_175306.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323221 |
AK029356.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125037 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P126447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23311892 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23311894 |
- |
- |