Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175324.3 | NP_780533.2 | 17 - 18 | 2057 | 67 | |
GenBank mRNA | AK077531.1 | - | 17 - 18 | 2058 | 67 | |
AK128998.1 | - | 17 - 18 | 2036 | 67 | ||
AK139918.1 | - | 17 - 18 | 2001 | 67 | ||
AK145561.1 | - | 17 - 18 | 2046 | 67 | ||
AK145834.1 | - | 17 - 18 | 2037 | 67 | ||
BC006042.1 | - | 7 - 8 | 733 | 67 | ||
BC032994.1 | - | 10 - 11 | 986 | 67 | ||
BC045199.1 | - | 17 - 18 | 2021 | 5133197 | 67 | |
BC110556.2 | - | 3 - 4 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |