Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175325.3 | NP_780534.1 | 10 - 11 | 784 | 78 | |
GenBank mRNA | AK039560.1 | - | 10 - 11 | 733 | 78 | |
AK089970.1 | - | 10 - 11 | 739 | 78 | ||
AK134734.1 | - | 10 - 11 | 737 | 78 | ||
BC055797.1 | - | 10 - 11 | 704 | 78 | ||
BC089507.1 | - | 10 - 11 | 718 | 78 | ||
BC092531.1 | - | 10 - 11 | 728 | 78 | ||
BC145771.1 | - | 10 - 11 | 746 | 40131074 | 78 | |
BC145773.1 | - | 10 - 11 | 746 | 40131076 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |