Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175561.4 | NP_780770.2 | 17 - 18 | 3448 | 77 | |
GenBank mRNA | AK087907.1 | - | 2 - 3 | 361 | 77 | |
AK220342.1 | - | 9 - 10 | 1010 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |