Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175669.3 | NP_783600.1 | 3 - 4 | 244 | 127 | |
GenBank mRNA | AK033761.1 | - | 3 - 4 | 246 | 127 | |
BC116704.1 | - | 3 - 4 | 239 | 40104936 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |