Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176838.2 | NP_789808.1 | 13 - 14 | 2104 | 77 | |
GenBank mRNA | AK088755.1 | - | 13 - 14 | 1974 | 77 | |
AK168488.1 | - | 12 - 13 | 1337 | 77 | ||
BC031444.1 | - | 13 - 14 | 2104 | 4222237 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |