Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176838.2 | NP_789808.1 | 1 - 2 | 404 | 83 | |
GenBank mRNA | AK088755.1 | - | 1 - 2 | 273 | 83 | |
AK168488.1 | - | 1 - 2 | 252 | 83 | ||
BC031444.1 | - | 1 - 2 | 404 | 4222237 | 83 | |
BC096581.1 | - | 1 - 2 | 244 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |